Merck
  • Evidence against a systemic arterial defect in patients with inflammatory bowel disease.
Evidence against a systemic arterial defect in patients with inflammatory bowel disease.
The Journal of surgical research (2014-05-14)
Luke A Neill, Christine M Saundry, Neil H Hyman, George C Wellman
ABSTRACT
Despite increasing interest in local microvascular alterations associated with inflammatory bowel disease (IBD), the potential contribution of a primary systemic vascular defect in the etiology of IBD is unknown. We compared reactivity of large diameter mesenteric arteries from segments affected by Crohn disease (CD) or ulcerative colitis (UC) to an uninvolved vascular bed in both IBD and control patients. Mesenteric and omental arteries were obtained from UC, CD, and non-IBD patients. Isometric arterial contractions were recorded in response to extracellular potassium (K(+)) and cumulative additions of norepinephrine (NE). In addition, relaxation in response to pinacidil, an activator of adenosine triphosphate-sensitive K(+) channels was examined. Contraction to K(+) and sensitivity to NE were not significantly different in arteries from CD, UC, and controls. Relaxation to pinacidil was also similar between groups. Potassium-induced contractions and sensitivity to NE and pinacidil were not significantly different in large diameter mesenteric and omental arteries obtained from IBD patients. Furthermore, there was no significant difference in the sensitivity to K(+), NE, and pinacidil between mesenteric and omental arteries of CD and UC patients and those from non-IBD patients. Our results suggest an underlying vascular defect systemic to CD or UC patients is unlikely to contribute to the etiology of IBD.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
L-Norepinephrine hydrochloride, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
DL-Norepinephrine hydrochloride, crystalline, ≥97% (TLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material