Merck
Photodegradation of the antihistamine cetirizine in natural waters.
Environmental toxicology and chemistry (2014-07-22)
Ralph N Mead, Seth Barefoot, John R Helms, Jeremy B Morgan, Robert J Kieber
ABSTRACT
The photodegradation rate of the anti-histamine cetirizine (Zyrtec®) was investigated in various water matrices. The average observed first-order photodegradation rate coefficient (kobs ), obtained by linear regression of the logarithmic-transformed cetirizine concentrations versus irradiation time in simulated sunlight, was 0.024 h(-1) (n = 6; standard deviation ± 0.004) in deionized water corresponding to a half-life of approximately 30 h. There was no statistical difference in the kobs of cetirizine photodegradation in coastal seawater compared with deionized water or deionized water amended with dissolved chromophoric organic matter. The quantum yield of cetirizine photodegradation decreased dramatically with increasing wavelength and decreasing energy of incoming radiation, with the average value ranging from 5.28 × 10(-4) to 6.40 × 10(-3) in the ultraviolet wavelength range (280-366 nm). The activation energy of cetirizine photodegradation was 10.3 kJ mol(-1) with an observed increase in cetirizine photodegradation as temperature increased. This is a significant environmental factor influencing half-life and an important consideration, given that cetirizine has been detected in wastewater and receiving waters from different locations globally.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, Deionized
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)