Merck
  • Enantioselective Mannich reaction of β-keto esters with aromatic and aliphatic imines using a cooperatively assisted bifunctional catalyst.
Enantioselective Mannich reaction of β-keto esters with aromatic and aliphatic imines using a cooperatively assisted bifunctional catalyst.
Organic letters (2014-09-18)
Antti J Neuvonen, Petri M Pihko
ABSTRACT
An efficient urea-enhanced thiourea catalyst enables the enantioselective Mannich reaction between β-keto esters and N-Boc-protected imines under mild conditions and minimal catalyst loading (1-3 mol %). Aliphatic and aromatic substituents are tolerated on both reaction partners, affording the products in good enantiomeric purity. The corresponding β-amino ketones can readily be accessed via decarboxylation without loss of enantiomeric purity.
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O