Merck
  • Florfenicol residues in rainbow trout after oral dosing in recirculating and flow-through culture systems.
Florfenicol residues in rainbow trout after oral dosing in recirculating and flow-through culture systems.
Journal of aquatic animal health (2014-10-17)
J R Meinertz, K R Hess, J A Bernardy, M P Gaikowski, M Whitsel, R G Endris
ABSTRACT
Aquaflor is a feed premix for fish containing the broad spectrum antibacterial agent florfenicol (FFC) incorporated at a ratio of 50% (w/w). To enhance the effectiveness of FFC for salmonids infected with certain isolates of Flavobacterium psychrophilum causing cold water disease, the FFC dose must be increased from the standard 10 mg·kg⁻¹ body weight (BW)·d⁻¹ for 10 consecutive days. A residue depletion study was conducted to determine whether FFC residues remaining in the fillet tissue after treating fish at an increased dose would be safe for human consumption. Groups of Rainbow Trout Oncorhynchus mykiss (total n = 144; weight range, 126-617 g) were treated with FFC at 20 mg·kg⁻¹ BW·d⁻¹ for 10 d in a flow-through system (FTS) and a recirculating aquaculture system (RAS) each with a water temperature of ∼13°C. The two-tank RAS included a nontreated tank containing 77 fish. Fish were taken from each tank (treated tank, n = 16; nontreated tank, n = 8) at 6, 12, 24, 48, 72, 120, 240, 360, and 480 h posttreatment. Florfenicol amine (FFA) concentrations (the FFC marker residue) in skin-on fillets from treated fish were greatest at 12 h posttreatment (11.58 μg/g) in the RAS and were greatest at 6 h posttreatment (11.09 μg/g) in the FTS. The half-lives for FFA in skin-on fillets from the RAS and FTS were 20.3 and 19.7 h, respectively. Assimilation of FFC residues in the fillets of nontreated fish sharing the RAS with FFC-treated fish was minimal. Florfenicol water concentrations peaked in the RAS-treated tank and nontreated tanks at 10 h (453 μg/L) and 11 h (442 μg/L) posttreatment, respectively. Monitoring of nitrite concentrations throughout the study indicated the nitrogen oxidation efficiency of the RAS biofilter was minimally impacted by the FFC treatment.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Supelco
Florfenicol amine, VETRANAL®, analytical standard
Supelco
Florfenicol, analytical standard, for drug analysis
Sigma-Aldrich
Thiamphenicol
Supelco
Water, for ultratrace analysis, ACS reagent
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, for TOC analysis
Supelco
Water
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Supelco
Thiamphenicol, analytical standard
Thiamphenicol, European Pharmacopoeia (EP) Reference Standard