Merck
  • Synergism of arsenic trioxide and MG132 in Raji cells attained by targeting BNIP3, autophagy, and mitochondria with low doses of valproic acid and vincristine.
Synergism of arsenic trioxide and MG132 in Raji cells attained by targeting BNIP3, autophagy, and mitochondria with low doses of valproic acid and vincristine.
European journal of cancer (Oxford, England : 1990) (2014-12-03)
Victoria Cavaliere, Tomás Lombardo, Susana N Costantino, Laura Kornblihtt, Elida M Alvarez, Guillermo A Blanco
ABSTRACT
We previously demonstrated that arsenic trioxide (ATO) and proteasome inhibitor MG132 synergistically induced cell death in promonocytic leukaemia cell line U937 but were antagonistic in Burkitt's lymphoma cell line Raji. Here we explore the role of autophagy, expression of BNIP3, and mitochondrial mass, in determining whether ATO and MG132 interaction can be shifted from antagonism to synergism in Raji cells. Treatment with ATO+MG132 increased the percentage of cells with collapsed mitochondrial membrane potential (MMP) in U937 cells, but had no effect in Raji cells. Mitochondria were found in cytoplasmic marginal location in U937 cells but at perinuclear location in Raji cells. ATO+MG132 increased mitochondrial mass in U937 cells but decreased it in Raji cells, while autophagy was increased in both cell lines. BNIP3 was expressed in U937 cells at cytoplasmic marginal locations and was hardly detected in Raji cells. Histone deacetylase (HDAC) inhibitor valproic acid (VPA) increased expression of BNIP3 in Raji cells at perinuclear locations. However antagonism between ATO and MG132 was increased in the presence of low doses of VPA. Addition of vincristine (VCR) blocked autophagy, while VPA+VCR treatment of Raji cells at sub-cytotoxic doses caused BNIP3 and mitochondria to redistribute to cytoplasmic peripheral location and increased mitochondrial mass. ATO+MG132 in the presence of subcytotoxic doses of VPA+VCR caused collapse of MMP in Raji cells, while interaction between ATO and MG132 shifted from antagonism to synergism. We conclude that synergism between ATO and MG132 was attained in Raji cells by disruption of the perinuclear mitochondrial cluster, blockage of selective autophagy of mitochondria (mitophagy) by VCR, increased mitochondrial mass, and upregulation of BNIP3 by VPA.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)