Merck
  • Photochemistry. Chemiexcitation of melanin derivatives induces DNA photoproducts long after UV exposure.
Photochemistry. Chemiexcitation of melanin derivatives induces DNA photoproducts long after UV exposure.
Science (New York, N.Y.) (2015-02-24)
Sanjay Premi, Silvia Wallisch, Camila M Mano, Adam B Weiner, Antonella Bacchiocchi, Kazumasa Wakamatsu, Etelvino J H Bechara, Ruth Halaban, Thierry Douki, Douglas E Brash
ABSTRACT
Mutations in sunlight-induced melanoma arise from cyclobutane pyrimidine dimers (CPDs), DNA photoproducts that are typically created picoseconds after an ultraviolet (UV) photon is absorbed at thymine or cytosine. We found that in melanocytes, CPDs are generated for >3 hours after exposure to UVA, a major component of the radiation in sunlight and in tanning beds. These "dark CPDs" constitute the majority of CPDs and include the cytosine-containing CPDs that initiate UV-signature C→T mutations. Dark CPDs arise when UV-induced reactive oxygen and nitrogen species combine to excite an electron in fragments of the pigment melanin. This creates a quantum triplet state that has the energy of a UV photon but induces CPDs by energy transfer to DNA in a radiation-independent manner. Melanin may thus be carcinogenic as well as protective against cancer. These findings also validate the long-standing suggestion that chemically generated excited electronic states are relevant to mammalian biology.
Product Description
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Cytosine, ≥99%
Supelco
Thymine(Zidovudine Related Compound C), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard