Merck
  • Development of a time-resolved fluorescence probe for evaluation of competitive binding to the cholecystokinin 2 receptor.
Development of a time-resolved fluorescence probe for evaluation of competitive binding to the cholecystokinin 2 receptor.
Bioorganic & medicinal chemistry (2015-03-15)
N G R Dayan Elshan, Thanuja Jayasundera, Craig S Weber, Ronald M Lynch, Eugene A Mash
ABSTRACT
The synthesis, characterization, and use of Eu-DTPA-PEGO-Trp-Nle-Asp-Phe-NH2 (Eu-DTPA-PEGO-CCK4), a luminescent probe targeted to cholecystokinin 2 receptor (CCK2R, aka CCKBR), are described. The probe was prepared by solid phase synthesis. A Kd value of 17±2nM was determined by means of saturation binding assays using HEK-293 cells that overexpress CCK2R. The probe was then used in competitive binding assays against Ac-CCK4 and three new trivalent CCK4 compounds. Repeatable and reproducible binding assay results were obtained. Given its ease of synthesis, purification, receptor binding properties, and utility in competitive binding assays, Eu-DTPA-PEGO-CCK4 could become a standard tool for high-throughput screening of compounds in development targeted to cholecystokinin receptors.
Sigma-Aldrich
Thyrotropin releasing hormone, ≥98% (HPLC), powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%