Merck
  • Quantifying protein measurands by peptide measurements: where do errors arise?
Quantifying protein measurands by peptide measurements: where do errors arise?
Journal of proteome research (2014-12-17)
Irene van den Broek, Fred P H T M Romijn, Nico P M Smit, Arnoud van der Laarse, Jan W Drijfhout, Yuri E M van der Burgt, Christa M Cobbaert
ABSTRACT
Clinically actionable quantification of protein biomarkers by mass spectrometry (MS) requires analytical performance in concordance with quality specifications for diagnostic tests. Laboratory-developed tests should, therefore, be validated in accordance with EN ISO 15189:2012 guidelines for medical laboratories to demonstrate competence and traceability along the entire workflow, including the selected standardization strategy and the phases before, during, and after proteolysis. In this study, bias and imprecision of a previously developed MS method for quantification of serum apolipoproteins A-I (Apo A-I) and B (Apo B) were thoroughly validated according to Clinical and Laboratory Standards Institute (CLSI) guidelines EP15-A2 and EP09-A3, using 100 patient sera and either stable-isotope labeled (SIL) peptides or SIL-Apo A-I as internal standard. The systematic overview of error components assigned sample preparation before the first 4 h of proteolysis as major source (∼85%) of within-sample imprecision without external calibration. No improvement in imprecision was observed with the use of SIL-Apo A-I instead of SIL-peptides. On the contrary, when the use of SIL-Apo A-I was combined with external calibration, imprecision improved significantly (from ∼9% to ∼6%) as a result of the normalization for matrix effects on linearity. A between-sample validation of bias in 100 patient sera further supported the presence of matrix effects on digestion completeness and additionally demonstrated specimen-specific biases associated with modified peptide sequences or alterations in protease activity. In conclusion, the presented overview of bias and imprecision components contributes to a better understanding of the sources of errors in MS-based protein quantification and provides valuable recommendations to assess and control analytical quality in concordance with the requirements for clinical use.
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
SAFC
Iodoacetamide
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)