Merck
  • Upregulated Annexin A1 promotes cellular invasion in triple-negative breast cancer.
Upregulated Annexin A1 promotes cellular invasion in triple-negative breast cancer.
Oncology reports (2015-01-17)
Maiko Okano, Kensuke Kumamoto, Motonobu Saito, Hisashi Onozawa, Katsuharu Saito, Noriko Abe, Tohru Ohtake, Seiichi Takenoshita
ABSTRACT
Annexin A1 (ANXA1) is a calcium-dependent phospholipid-linked protein, involved in anti-inflammatory effects, regulation of cellular differentiation, proliferation and apoptosis. While many studies have investigated the ANXA1 expression in various tumor types, the role of ANXA1 is not fully understood. Therefore, in the present study, we evaluated the ANXA1 expression in 211 breast cancer patients and compared the levels with clinicopathological factors. ANXA1 was positively expressed in 31 (14.7%) of the 211 cases in our cohort, and these positive cases were associated with triple-negative breast cancer (TNBC) (P=0.007) and venous invasion (P=0.028). The in vitro cell experiment found that the MDA-MB-231 cell line, which is a TNBC cell line, highly expressed ANXA1. Using this cell line, the functional role of ANXA1 in breast cancer was revealed and the knockdown of ANXA1 by specific siRNA demonstrated a significant reduction in cellular invasion. Further experiments indicated that ANXA1 was induced by hypoxia with hypoxia-inducible factor-1α induction. These results suggested that ANXA1, which enhanced breast cancer invasion and metastasis under hypoxia, were significantly associated with the worst patient outcome. This is particularly noted in TNBC, the group of breast cancer with the worst outcome for which new therapeutic implications are required.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
SAFC
BIS-TRIS
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard