Merck
  • Development of phosphatase inhibitor-1 peptides acting as indirect activators of phosphatase 1.
Development of phosphatase inhibitor-1 peptides acting as indirect activators of phosphatase 1.
Naunyn-Schmiedeberg's archives of pharmacology (2014-11-25)
Hannieh Sotoud, Uwe Borgmeyer, Christian Schulze, Ali El-Armouche, Thomas Eschenhagen
ABSTRACT
Phosphatase inhibitor-1 (I-1) inhibits the catalytic subunit of protein phosphatase type 1 (PP1c) in its protein kinase A (PKA)-phosphorylated form (I-1(P)). It thereby amplifies PKA signaling, which, in the heart, mediates both beneficial (acute) and adverse (chronic) effects of catecholamines. Genetic deletion of I-1 was associated with protection against catecholamine toxicity, making the PP1c-I-1(P) complex a potential therapeutic target for chronic heart disease. Here, we sought to define targetable interaction sites of I-1 and PP1c, concentrating on the N-terminal domain of I-1 which includes the PP1c binding motif ((9)KIQF(12)) as well as a poly-Arg stretch. Substitution of (9)KIQ(11) residues for analogous amino acids, (9)RLN(11), resulted in doubling of the IC50 values, deletion of (9)KIQF(12) prevented I-1 PKA-dependent phosphorylation and thus activation. Mutation of the Arg residues preceding the PKA phosphorylation site (Thr35) to Ala (R/A(30-33)) abolished I-1 phosphorylation and its binding to and inhibition of PP1c. A series of synthetic peptides (4-11 residues) indicated that the KIQF motif as well as the surrounding anchoring residues was essential for interfering with the inhibitory effect of I-1(P) on PP1c, whereas the four Arg residues were not. Unexpectedly, the most effective nonapeptide (SPRKIQFTV) also antagonized the inhibitory effect of the non-conditional PP1 inhibitor-2 with similar affinity. Incubation of neonatal rat cardiac myocytes with a poly-Arg-modified SPRKIQFTV (10 μM) reduced catecholamine-induced phosphorylation of phospholamban, a well-known PKA downstream target sensitive to PP1c. Our data reiterate the importance of the KIQF motif and provide a tool for antagonizing I-1 inhibitory effects on PP1c, i.e., activating PP1 in vivo.
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Threonine
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri