Merck
  • Improved quality of surgical field during endoscopic sinus surgery after clonidine premedication--a pilot study.
Improved quality of surgical field during endoscopic sinus surgery after clonidine premedication--a pilot study.
International forum of allergy & rhinology (2014-03-05)
Katarzyna Wawrzyniak, Pawel K Burduk, Jacek B Cywinski, Krzysztof Kusza, Wojciech Kazmierczak
ABSTRACT
Inadequate surgical field visualization due to intraoperative bleeding during endoscopic sinus surgery (ESS) can cause major complications. The aim of this prospective study was to compare the effect of preoperative administration of clonidine and melatonin on the quality of the surgical field visualization and selected aspects of presurgical premedication. Twenty-six patients undergoing ESS for chronic sinusitis and polyp removal were enrolled and randomly assigned to receive either oral clonidine or melatonin as preoperative premedication. During the operation, the quality of the surgical field was assessed and graded using the scale proposed by Boezaart. The evaluations were done at 15, 30, and 60 minutes after incision. The effect of this premedication choice on the intraoperative and postoperative hemodynamic profile was examined in each group. Anxiolytic effects of both premedication agents were assessed using the visual analogue scale for anxiety (VAS-A). Propofol dose and induction time (the time for bispectral index [BIS] to reach 45), anesthesia and surgical procedures time, and recovery time (the time for BIS to reach 90) were assessed in both groups as well. The quality of the surgical field was consistently better in 2 of 3 time points in the clonidine group. Perioperative mean arterial pressure and intraoperative heart rate had a more favorable profile in patients premedicated with clonidine. There were no differences in other measured parameters between groups. Premedication with clonidine before ESS provides better quality of surgical field and more favorable hemodynamic profile as compared to melatonin.
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Propofol, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard