Merck
  • Novel solvates M(BH₄)₃S(CH₃)₂ and properties of halide-free M(BH₄)₃ (M = Y or Gd).
Novel solvates M(BH₄)₃S(CH₃)₂ and properties of halide-free M(BH₄)₃ (M = Y or Gd).
Dalton transactions (Cambridge, England : 2003) (2014-07-26)
Morten B Ley, Mark Paskevicius, Pascal Schouwink, Bo Richter, Drew A Sheppard, Craig E Buckley, Torben R Jensen
ABSTRACT
Rare earth metal borohydrides have been proposed as materials for solid-state hydrogen storage because of their reasonably low temperature of decomposition. New synthesis methods, which provide halide-free yttrium and gadolinium borohydride, are presented using dimethyl sulfide and new solvates as intermediates. The solvates M(BH4)3S(CH3)2 (M = Y or Gd) are transformed to α-Y(BH4)3 or Gd(BH4)3 at ~140 °C as verified by thermal analysis. The monoclinic structure of Y(BH4)3S(CH3)2, space group P2₁/c, a = 5.52621(8), b = 22.3255(3), c = 8.0626(1) Å and β = 100.408(1)°, is solved from synchrotron radiation powder X-ray diffraction data and consists of buckled layers of slightly distorted octahedrons of yttrium atoms coordinated to five borohydride groups and one dimethyl sulfide group. Significant hydrogen loss is observed from Y(BH4)3 below 300 °C and rehydrogenation at 300 °C and p(H2) = 1550 bar does not result in the reformation of Y(BH4)3, but instead yields YH3. Moreover, composites systems Y(BH4)3-LiBH4 1 : 1 and Y(BH4)3-LiCl 1 : 1 prepared from as-synthesised Y(BH4)3 are shown to melt at 190 and 220 °C, respectively.
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Supelco
Toluene, analytical standard
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
Borane dimethyl sulfide complex
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Borane dimethyl sulfide complex solution, 2.0 M in THF
Supelco
Electrolyte solution, nonaqueous, LiCl in ethanol (saturated)
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Borane dimethyl sulfide complex solution, 1.0 M in 2-methyltetrahydrofuran
Supelco
Dimethyl sulfide, analytical standard
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%