Merck
  • The comparison of dispersive solid phase extraction and multi-plug filtration cleanup method based on multi-walled carbon nanotubes for pesticides multi-residue analysis by liquid chromatography tandem mass spectrometry.
The comparison of dispersive solid phase extraction and multi-plug filtration cleanup method based on multi-walled carbon nanotubes for pesticides multi-residue analysis by liquid chromatography tandem mass spectrometry.
Journal of chromatography. A (2015-02-11)
Yuhong Qin, Pengyue Zhao, Sufang Fan, Yongtao Han, Yanjie Li, Nan Zou, Shuangyu Song, Yuan Zhang, Fangbing Li, Xuesheng Li, Canping Pan
ABSTRACT
In this study, dispersive-Solid Phase Extraction (d-SPE) cleanup and multi-plug filtration cleanup (m-PFC) methods were compared for 25 representative pesticides in six matrices (wheat, spinach, carrot, apple, citrus and peanut) by QuEChERS-LC-ESI-MS/MS detection. The type of sorbents in dispersive-SPE (d-SPE) was optimized for the above matrices. Multi-walled carbon nanotubes (MWCNTs), which mixed other materials like PSA (Primary Secondary Amines), GCB (Graphitized Carbon Black) and C18 (Octadecyl-silica), showed brilliant cleanup performance in multi residue monitoring (MRM) pesticide residue analysis. Cleanup effects with d-SPE and m-PFC methods were examined. When spiked at 3 concentration levels of 10, 100, 500 μg/kg in above matrices, for both d-SPE and m-PFC methods, the recoveries ranged from 70 to 110% with relative standard deviations (RSDs) lower than 20%. Limits of quantification (LOQs) for both cleanup methods ranged from 1 to 25 μg/kg. Matrix-matched calibrations were performed with the coefficients of determination more than 0.99 between concentration levels of 10-1000 μg/kg. It was found that m-PFC was more convenient and effective than d-SPE with the same sorbents, due to the increased contact time and contact area between the extracts and compressed sorbents. The study demonstrated that m-PFC method could be used as a rapid, convenient and high-throughput cleanup method for analysis of pesticide residues.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%