Merck
  • Vitrification alters rabbit foetal placenta at transcriptomic and proteomic level.
Vitrification alters rabbit foetal placenta at transcriptomic and proteomic level.
Reproduction (Cambridge, England) (2014-02-19)
M D Saenz-de-Juano, F Marco-Jimenez, B Schmaltz-Panneau, E Jimenez-Trigos, M P Viudes-de-Castro, D S Peñaranda, L Jouneau, J Lecardonnel, R Lavara, C Naturil-Alfonso, V Duranthon, J S Vicente
ABSTRACT
Although numerous studies have demonstrated that cryopreservation alters gene expression, less is known about those embryos that implanted successfully and continued in gestation. To raise the question of the neutrality of this technique, we examine the effects of vitrification through gestation in rabbit before and after the implantation. We monitored the distribution of losses of 569 vitrified morulae, observing that embryos which reach the last pre-implantatory stage are able to implant. However, we found that not all implanted embryos had the ability to continue with their gestation. The results reveal that vitrification decreased foetus and maternal placenta weights at mid-gestation, but led to a higher offspring birth weight. A novel finding is that while no differences in gene expression were detected in pre-implantatory embryos at day 6, vitrification affects a gene and protein expression in the placenta at day 14. Our results for first time reveal strong evidence of modifications in implanted embryos subjected to vitrification, suggesting that the crucial step that vitrified embryos must overcome is the placenta formation. On the basis of these findings, our work leaves the question open as to whether the effects we observed that cause vitrification during foetal development could give rise to some type of physiological or metabolic alteration in adulthood.
Product Description
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, natural, 97%, FG
Pricing and availability is not currently available.
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Pricing and availability is not currently available.
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Pricing and availability is not currently available.
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99%
Pricing and availability is not currently available.
Supelco
Phenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Pricing and availability is not currently available.
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Xylazine, ≥99%
Pricing and availability is not currently available.