Merck
  • Physico-chemical and toxicological characterization of iron-containing albumin nanoparticles as platforms for medical imaging.
Physico-chemical and toxicological characterization of iron-containing albumin nanoparticles as platforms for medical imaging.
Journal of controlled release : official journal of the Controlled Release Society (2014-09-01)
Ina Rosenberger, Christian Schmithals, Jennifer Vandooren, Silvia Bianchessi, Paolo Milani, Erica Locatelli, Liron L Israel, Frank Hübner, Michela Matteoli, Jean-Paul Lellouche, Mauro Comes Franchini, Lorena Passoni, Eugenio Scanziani, Ghislain Opdenakker, Albrecht Piiper, Jörg Kreuter
ABSTRACT
Iron oxide-containing magnetic nanoparticles (MNPs) have certain advantages over currently used contrast agents for tumor imaging by magnetic resonance imaging (MRI) as they offer the possibility of functionalization with ligands and tracers. Functionalized MNPs also may be used for targeted tumor therapy. In the current study nanoparticles (NPs) consisting of recombinant human serum albumin (rHSA) with incorporated hydrophilic (NH4)2Ce(IV)(NO3)6-γ-Fe2O3 particles (CAN maghemite particles) for medical imaging were produced and characterized. For this purpose CAN maghemite particles were incorporated into an rHSA matrix to yield rHSA-NPs. The resulting NPs were analyzed by transmission electron microscopy, photon correlation spectroscopy, and atomic absorption. The sizes of the manufactured NP were 170 ± 10 nm, and the zeta-potential was -50 ± 3 mV. The NPs remained stable when stored after lyophilization with sucrose 3% [w/v] as a cryoprotector. They showed pro-inflammatory properties without cell and animal toxicity in vivo and were highly contrasting in MRI. In conclusion, this report introduces novel rHSA NP with favorable properties containing iron oxide for detection by MRI.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)