Merck
  • A novel approach to enhance the mucoadhesion of lipid drug nanocarriers for improved drug delivery to the buccal mucosa.
A novel approach to enhance the mucoadhesion of lipid drug nanocarriers for improved drug delivery to the buccal mucosa.
International journal of pharmaceutics (2014-06-01)
Joanne D Du, Qingtao Liu, Stefan Salentinig, Tri-Hung Nguyen, Ben J Boyd
ABSTRACT
Targeted drug delivery to the buccal mucosa offers distinct advantages over oral delivery to the gastrointestinal tract including by-passing hepatic first-pass metabolism. However, the buccal route is often limited by low bioavailability, low drug loading and reduced residence time due to salivary excretion and clearance. To overcome these limitations, a novel mucoadhesive formulation based on liquid crystalline nanoparticles was designed. Utilising a pH induced in situ transition from a stable vesicle formulation to dispersed inverse hexagonal phase nanoparticles (hexosomes) enhanced adsorption onto the mucosal surface was enabled. Firstly, the phase behaviour of the amphiphilic lipid phytantriol (PHY) and oleic acid (OA) was assessed from pH 2-9 using small-angle X-ray scattering (SAXS) and cryo-transmission electron microscopy (cryo-TEM) to determine the appropriate composition for the vesicle to hexosome transition. The colloidal stability of the formulation was determined using turbidity studies. Dispersions comprising 30% w/w OA in PHY were able to form stable vesicles at pH 8 and transition to hexosomes when exposed to pH<7 (as encountered on the buccal mucosal surface). Subsequent ex vivo studies utilising excised porcine buccal tissue indicated significant retention of the in situ-formed PHY/OA hexosomes when compared to control DOPC vesicles (p<0.005), confirmed independently using confocal fluorescence microscopy, radioactive scintillation counting and HPLC analysis for incorporated drug. Thus, a novel approach providing a stable vesicle formulation, with in situ transformation to mucoadhesive hexosomes has been identified with the potential to enhance drug delivery to mucosal surfaces.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Supelco
Triamcinolone acetonide, analytical standard
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis