Merck
  • The impact of early intra-articular administration of interleukin-1 receptor antagonist on lubricin metabolism and cartilage degeneration in an anterior cruciate ligament transection model.
The impact of early intra-articular administration of interleukin-1 receptor antagonist on lubricin metabolism and cartilage degeneration in an anterior cruciate ligament transection model.
Osteoarthritis and cartilage (2014-09-16)
K A Elsaid, L Zhang, Z Shaman, C Patel, T A Schmidt, G D Jay
ABSTRACT
Study the impact of intra-articular interleukin-1 receptor antagonist (IL-1 ra) treatment on lubricin biosynthesis following anterior cruciate ligament transection (ACLT) in the rat and evaluate the effect of combined IL-1 ra and recombinant human lubricin (rhPRG4) treatments on chondrocyte apoptosis. ACLT was performed in male Lewis rats. Treatments included IL-1 ra or vehicle (n = 36 in each group). IL-1 ra intra-articular dosing was performed on days 1, 3, 5 and 7 following ACLT using Anakinra (150 mg/ml; 40 μl). At 3 and 5 weeks, animals were sacrificed and RNA was isolated. Histological analyses included Safranin O and H&E. Lubricin synovial fluid (SF) lavage concentrations were determined at 5 weeks. ACLT animals were treated with a single injection of vehicle, IL-1 ra (75 mg/ml; 40 μl), rhPRG4 (200 μg/ml; 40 μl), or IL-1 ra + rhPRG4 (75 mg/ml + 200 μg/ml; 40 μl) (n = 6 in each group) on day 7 following ACLT and cartilage was probed for cleaved caspase-3 at 5 weeks. IL-1 ra treatment improved lubricin expression (P < 0.001) and lubricin SF lavage concentrations in the IL-1 ra group was higher (P = 0.005) than the vehicle. IL-1 ra treatment reduced cartilage and synovial scores (P < 0.001) compared to vehicle. IL-1 ra and rhPRG4 acted synergistically to reduce caspase-3 positive chondrocytes (P < 0.001) compared to individual treatments. IL-1 ra treatment preserved lubricin following ACLT and a combined treatment of IL-1 ra + rhPRG4 may act synergistically to reduce cartilage catabolism.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
SAFC
L-Glutamine
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
L-Glutamine
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland