Merck
  • PfRON3 is an erythrocyte-binding protein and a potential blood-stage vaccine candidate antigen.
PfRON3 is an erythrocyte-binding protein and a potential blood-stage vaccine candidate antigen.
Malaria journal (2014-12-17)
Xin Zhao, Zhiguang Chang, Zhiwei Tu, Shengchao Yu, Xiaoyan Wei, Jianhua Zhou, Huijun Lu, Ning Jiang, Qijun Chen
ABSTRACT
Erythrocyte invasion by merozoites is an essential step in Plasmodium falciparum infection and leads to subsequent disease pathology. Proteins both on the merozoite surface and secreted from the apical organelles (micronemes, rhoptries and dense granules) mediate the invasion of erythrocytes; some of the molecules have been regarded as targets in the development of an anti-malaria vaccine. Recently, a subgroup of rhoptry neck proteins (PfRON2, PfRON4 and PfRON5) associated with the microneme protein apical membrane antigen AMA1 has been described as components of the moving junction complex that assists merozoite invasion into erythrocytes. However, unlike PfRON2, PfRON4 and PfRON5, the latest study suggested that PfRON3 might be located in the rhoptry bulb and participates in a novel PfRON complex (PfRON2, 3 and 4), but does not form a complex with AMA1. Additionally, the full-length PfRON3 protein possesses three transmembrane regions at the N-terminus, which is highly conserved among RON3 orthologues in the genus Plasmodium, Toxoplasma gondii and Eimeria tenella. Overall, these findings suggest that PfRON3 may play an important role in merozoite invasion into erythrocytes. PfRON3 was primarily expressed during the late trophozoite stage, with a peak in transcription levels at 40 hours post-invasion. The subcellular localization of PfRON3 was confirmed that it is a merozoite rhoptry bulb protein. Additionally, the recombinant form of PfRON3 protein bound to the erythrocyte and was recognized by sera collected from malaria endemic areas in Africa, and anti-PfRON3 antibodies significantly inhibited merozoite invasion into erythrocytes. The expression of PfRON3 was analysed via real-time quantitative PCR, and the recombinant PfRON3 proteins were generated with an Escherichia coli expression system. The subcellular localization of PfRON3 was assessed with immunoelectron microscopy and immunofluorescence assay (IFA). The recognition PfRON3 by malaria immune sera was analysed with an enzyme-linked immunosorbent assay (ELISA). Erythrocyte-binding assays were performed using recombinant PfRON3 proteins and invasion inhibition assays were carried out with PfRON3-specific antibodies. This study confirmed that PfRON3 is a rhoptry protein with an erythrocyte-binding property, which is likely associated red blood cell invasion. PfRON3 is a potential vaccine candidate.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents