Merck
  • In vitro effects of the organophosphorus pesticide malathion on the reactivity of rat aorta.
In vitro effects of the organophosphorus pesticide malathion on the reactivity of rat aorta.
Pharmacology (2014-10-11)
Marco Tulio Menezes de Carvalho, Andrea Carla Celotto, Agnes Afrodite Sumarelli Albuquerque, Luciana Garros Ferreira, Verena Kise Capellini, Ana Paula Cassiano Silveira, Tales Rubens de Nadai, Paulo Roberto Barbosa Evora
ABSTRACT
There is a remarkable paucity of studies analyzing the role of the endothelium-derived relaxing factors on the vascular effects of organophosphates. This study was carried out to evaluate the vascular effects of malathion and the role of nitric oxide (NO) and prostacyclin (PGI2). Vascular reactivity measuring isometric forces in vitro ('organ chambers') and flow cytometry (cells loaded with DAF-FM DA) were used. In rat thoracic aorta segments contracted with phenylephrine (Phe) (10(-7) mol/l), malathion (10(-10) to 10(-5) mol/l) induced concentration-dependent relaxation in arteries with intact endothelium (n = 7; p < 0.05). Malathion-mediated relaxation was blocked by N-nitro-L-arginine methyl ester (L-NAME; 10(-4) mol/l), a nonspecific NO synthase inhibitor, and/or indomethacin (10(-5) mol/l), a nonspecific cyclooxygenase inhibitor (n = 10, p < 0.05). In thoracic aorta rings, with and without endothelium, Phe (10(-10) to 10(-5) mol/l) evoked concentration-dependent contraction, which was reduced in the presence of malathion. In rings with or without endothelium, incubated with malathion, L-NAME and indomethacin, the Phe-induced contraction was restored. The role of NO was confirmed using flow cytometry. Malathion evokes endothelium-dependent relaxation through the M1 muscarinic receptor, since this relaxation was clearly blocked by atropine (M1 and M2 blocker) and pirenzepine (M1 blocker), but was less blocked by gallamine (M2 blocker) or 4-DAMP (M3 blocker). These findings suggest that the organophosphate compound effects on vascular reactivity depend of NO and PGI2.
Product Description
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-DAMP, ≥98% (HPLC)
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Malathion, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Atropine, European Pharmacopoeia (EP) Reference Standard
USP
Malathion, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Malathion, PESTANAL®, analytical standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture