Merck
  • A multifunctional polymeric nanotheranostic system delivers doxorubicin and imaging agents across the blood-brain barrier targeting brain metastases of breast cancer.
A multifunctional polymeric nanotheranostic system delivers doxorubicin and imaging agents across the blood-brain barrier targeting brain metastases of breast cancer.
ACS nano (2014-10-14)
Jason Li, Ping Cai, Alireza Shalviri, Jeffrey T Henderson, Chunsheng He, Warren D Foltz, Preethy Prasad, Peter M Brodersen, Yonghong Chen, Ralph DaCosta, Andrew Michael Rauth, Xiao Yu Wu
ABSTRACT
Metastatic brain cancers, in particular cancers with multiple lesions, are one of the most difficult malignancies to treat owing to their location and aggressiveness. Chemotherapy for brain metastases offers some hope. However, its efficacy is severely limited as most chemotherapeutic agents are incapable of crossing the blood-brain barrier (BBB) efficiently. Thus, a multifunctional nanotheranostic system based on poly(methacrylic acid)-polysorbate 80-grafted-starch was designed herein for the delivery of BBB-impermeable imaging and therapeutic agents to brain metastases of breast cancer. In vivo magnetic resonance imaging and confocal fluorescence microscopy were used to confirm extravasation of gadolinium and dye-loaded nanoparticles from intact brain microvessels in healthy mice. The targetability of doxorubicin (Dox)-loaded nanoparticles to intracranially established brain metastases of breast cancer was evaluated using whole body and ex vivo fluorescence imaging of the brain. Coexistence of nanoparticles and Dox in brain metastatic lesions was further confirmed by histological and microscopic examination of dissected brain tissue. Immuno-histochemical staining for caspase-3 and terminal-deoxynucleotidyl transferase dUTP nick end labeling for DNA fragmentation in tumor-bearing brain sections revealed that Dox-loaded nanoparticles selectively induced cancer cell apoptosis 24 h post-injection, while sparing normal brain cells from harm. Such effects were not observed in the mice treated with free Dox. Treatment with Dox-loaded nanoparticles significantly inhibited brain tumor growth compared to free Dox at the same dose as assessed by in vivo bioluminescence imaging of the brain metastases. These findings suggest that the multifunctional nanoparticles are promising for the treatment of brain metastases.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) chloride hexahydrate, 99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Gadolinium(III) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Fluoresceinamine, isomer I
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%