Merck
  • Impaired expression of type I and type II interferon receptors in HCV-associated chronic liver disease and liver cirrhosis.
Impaired expression of type I and type II interferon receptors in HCV-associated chronic liver disease and liver cirrhosis.
PloS one (2014-09-30)
Partha K Chandra, Feyza Gunduz, Sidhartha Hazari, Ramazan Kurt, Rajesh Panigrahi, Bret Poat, David Bruce, Ari J Cohen, Humberto E Bohorquez, Humberto E Behorquez, Ian Carmody, George Loss, Luis A Balart, Tong Wu, Srikanta Dash
ABSTRACT
Chronic Hepatitis C Virus (HCV)-infected patients with liver cirrhosis (LC) respond poorly to interferon-alpha (IFN-α) and ribavirin (RBV) combination therapy, but the reason for this is unclear. We previously reported that HCV-infection induces endoplasmic reticulum (ER) stress and autophagy response that selectively down regulates the type I IFN-α receptor-1 (IFNAR1) and RBV transporters (CNT1 and ENT1), leading to IFN-α/RBV resistance. The goal of this study is to verify whether an increase in ER stress and autophagy response is also associated with the reduced expression of IFNAR1 and RBV transporters in chronic HCV-infected patients. Primary human hepatocytes (PHH) were infected with cell culture grown HCV particles (JFH-ΔV3-Rluc). HCV replication was confirmed by the detection of viral RNA by RT-qPCR and HCV-core protein by Western blotting. The ER stress and autophagy response and expression of IFN receptors and RBV transporters in HCV infected PHH and liver tissues derived from patients were measured by Western blotting. HCV infection of PHH showed impaired expression of IFNAR1, IFNγR1 (Type II IFN receptor) and RBV transporters but not IL10Rβ (Type III IFN-λ receptor). ER stress markers (BiP, IRE1α and peIF2α) and autophagy response (LC3II, Beclin 1 and ATG5) were induced in HCV infected chronic liver disease (CLD) and LC patients. Liver biopsies (CLD) show a 50% reduced expression of IFNAR1 and RBV transporters. Furthermore, the expression of IFNAR1 and RBV transporters was impaired in almost all LC patients. HCV infection induces ER stress and autophagy response in infected PHH and chronically infected liver tissues. The expression of IFNAR1, IFNγR1 and RBV transporters were significantly impaired in CLD and cirrhotic livers. Our study provides a potential explanation for the reduced response rate of IFN-α and RBV combination therapy in HCV infected patients with liver cirrhosis.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard