Merck
  • Cysteine sulfinic acid decarboxylase regulation: A role for farnesoid X receptor and small heterodimer partner in murine hepatic taurine metabolism.
Cysteine sulfinic acid decarboxylase regulation: A role for farnesoid X receptor and small heterodimer partner in murine hepatic taurine metabolism.
Hepatology research : the official journal of the Japan Society of Hepatology (2013-09-17)
Thomas A Kerr, Yuri Matsumoto, Hitoshi Matsumoto, Yan Xie, Lawrence L Hirschberger, Martha H Stipanuk, Sayeepriyadarshini Anakk, David D Moore, Mitsuhiro Watanabe, Susan Kennedy, Nicholas O Davidson
ABSTRACT
Bile acid synthesis is regulated by nuclear receptors including farnesoid X receptor (FXR) and small heterodimer partner (SHP), and by fibroblast growth factor 15/19 (FGF15/19). We hypothesized that hepatic cysteine sulfinic acid decarboxylase (CSAD) (a key enzyme in taurine synthesis) is regulated by bile acids (BA). The aim of this study was to investigate CSAD regulation by BA dependent regulatory mechanisms. Mice were fed a control diet or a diet supplemented with either 0.5% cholate or 2% cholestyramine. To study BA dependent pathways, we utilized GW4064 (FXR agonist), FGF19 or T-0901317 (liver X receptor [LXR] agonist) and Shp-/- mice. Tissue mRNA was determined by quantitative reverse transcription polymerase chain reaction. Amino acids were measured by high-performance liquid chromatography. Mice supplemented with dietary cholate exhibited reduced hepatic CSAD mRNA while those receiving cholestyramine exhibited increased mRNA. Activation of FXR suppressed CSAD mRNA expression whereas CSAD expression was increased in Shp-/- mice. Hepatic hypotaurine concentration (the product of CSAD) was higher in Shp-/- mice with a corresponding increase in serum taurine conjugated BA. FGF19 administration suppressed hepatic cholesterol 7-α-hydroxylase (CYP7A1) mRNA but did not change CSAD mRNA expression. LXR activation induced CYP7A1 mRNA yet failed to induce CSAD mRNA expression. BA regulate CSAD mRNA expression in a feedback fashion via mechanisms involving SHP and FXR but not FGF15/19 or LXR. These findings implicate BA as regulators of CSAD mRNA via mechanisms shared with CYP7A1.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
GW4064, ≥97% (HPLC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri