Merck
Microdroplet fusion mass spectrometry for fast reaction kinetics.
Proceedings of the National Academy of Sciences of the United States of America (2015-03-17)
Jae Kyoo Lee, Samuel Kim, Hong Gil Nam, Richard N Zare
ABSTRACT
We investigated the fusion of high-speed liquid droplets as a way to record the kinetics of liquid-phase chemical reactions on the order of microseconds. Two streams of micrometer-size droplets collide with one another. The droplets that fused (13 μm in diameter) at the intersection of the two streams entered the heated capillary inlet of a mass spectrometer. The mass spectrum was recorded as a function of the distance x between the mass spectrometer inlet and the droplet fusion center. Fused droplet trajectories were imaged with a high-speed camera, revealing that the droplet fusion occurred approximately within a 500-μm radius from the droplet fusion center and both the size and the speed of the fused droplets remained relatively constant as they traveled from the droplet fusion center to the mass spectrometer inlet. Evidence is presented that the reaction effectively stops upon entering the heated inlet of the mass spectrometer. Thus, the reaction time was proportional to x and could be measured and manipulated by controlling the distance x. Kinetic studies were carried out in fused water droplets for acid-induced unfolding of cytochrome c and hydrogen-deuterium exchange in bradykinin. The kinetics of the former revealed the slowing of the unfolding rates at the early stage of the reaction within 50 μs. The hydrogen-deuterium exchange revealed the existence of two distinct populations with fast and slow exchange rates. These studies demonstrated the power of this technique to detect reaction intermediates in fused liquid droplets with microsecond temporal resolution.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium 2,6-dichloroindophenolate hydrate, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.