Merck
  • Platelet mitochondrial DNA methylation: a potential new marker of cardiovascular disease.
Platelet mitochondrial DNA methylation: a potential new marker of cardiovascular disease.
Clinical epigenetics (2015-04-23)
Andrea A Baccarelli, Hyang-Min Byun
ABSTRACT
Platelets are critical in the etiology of cardiovascular disease (CVD), and the mitochondria in these cells serve as an energy source for platelet function. Epigenetic factors, especially DNA methylation, have been employed as markers of CVD. Unlike nuclear DNA methylation, mitochondrial DNA (mtDNA) methylation has not been widely studied, in part, due to debate about its existence and role. In this study, we examined platelet mtDNA methylation in relation to CVD. We measured mtDNA methylation in platelets by bisulfite-PCR pyrosequencing and examined associations of CVD with methylation in mitochondrial genes; cytochrome c oxidase (MT-CO1, MT-CO2, and MT-CO3); tRNA leucine 1 (MT-TL1); ATP synthase (MT-ATP6 and MT-ATP8); and NADH dehydrogenase (MT-MD5). We report that CVD patients have significantly higher mtDNA methylation than healthy controls in MT-CO1 (18.53%, P < 0.0001), MT-CO2 (3.33%, P = 0.0001), MT-CO3 (0.92%, P < 0.0001), and MT-TL1 (1.67%, P = 0.0001), which are involved in ATP synthesis. Platelet mtDNA methylation was not related with age, BMI, and race in this study. Our results suggest that platelet mtDNA methylation, which could serve as non-invasive and easy-to-obtain markers, may be implicated in the etiology of CVD.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Vinyl laurate, Wacker Chemie AG, ≥98% (GC)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Cytosine, ≥99%
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Vinyl laurate, ≥99.0% (GC)
Sigma-Aldrich
Thymidine, ≥99%