Merck
  • IL-27 alleviates the bleomycin-induced pulmonary fibrosis by regulating the Th17 cell differentiation.
IL-27 alleviates the bleomycin-induced pulmonary fibrosis by regulating the Th17 cell differentiation.
BMC pulmonary medicine (2015-04-19)
Zhaoxing Dong, Xin Lu, Yanni Yang, Tao Zhang, Yongxia Li, Yanlin Chai, Wen Lei, Changbo Li, Li Ai, Wenlin Tai
ABSTRACT
Interleukin-27 (IL-27) is a multifunctional cytokine with both pro-inflammatory and immunoregulatory functions. At present, the role of IL-27 in pulmonary fibrosis remains unknown. In this study, we observed the expression of IL-27/IL-27R in a mouse model of bleomycin (BLM)-induced pulmonary fibrosis. We verified the role of IL-27 using hematoxylin and eosin as well as Masson's staining methods and measuring the content of hydroxyproline as well as collagen I and III. We assessed the differentiation of T lymphocytes in the spleen and measured the concentration of cytokines in bronchoalveolar lavage fluid (BALF) and the expression level of relevant proteins in the JAK/STAT and TGF-ß/Smad signaling pathways in lung tissue. Increased IL-27 expression in BLM-induced pulmonary fibrosis was noted. IL-27 treatment may alleviate pulmonary fibrosis and increase the survival of mice. IL-27 inhibited the development of CD4(+) IL-17(+), CD4(+) IL-4(+) T, and CD4(+) Foxp3(+) cells and the secretion of IL-17, IL-4, IL-6, and TGF-ß. IL-27 induced the production of CD4(+) IL-10(+) and CD4(+) INF-γ(+) T cells. IL-27 decreased the levels of phosphorylated STAT1, STAT3, STAT5, Smad1, and Smad3 but increased the level of SOCS3. This study demonstrates that IL-27 potentially attenuates BLM-induced pulmonary fibrosis by regulating Th17 differentiation and cytokine secretion.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard