Merck
  • Human mesenchymal stem cell spheroids in fibrin hydrogels exhibit improved cell survival and potential for bone healing.
Human mesenchymal stem cell spheroids in fibrin hydrogels exhibit improved cell survival and potential for bone healing.
Cell and tissue research (2014-05-02)
Kaitlin C Murphy, Sophia Y Fang, J Kent Leach
ABSTRACT
Mesenchymal stem cells (MSCs) have great therapeutic potential for the repair of nonhealing bone defects, because of their proliferative capacity, multilineage potential, trophic factor secretion and lack of immunogenicity. However, a major challenge to the translation of cell-based therapies into clinical practice is ensuring their survival and function upon implantation into the defect site. We hypothesize that forming MSCs into more physiologic three-dimensional spheroids, rather than employing dissociated cells from two-dimensional monolayer culture, will enhance their survival when exposed to a harsh microenvironment but maintain their osteogenic potential. MSC spheroids were formed by using the hanging drop method with increasing cell numbers. Compared with larger spheroids, the smallest spheroids, which contained 15,000 cells, exhibited increased metabolic activity, reduced apoptosis and the most uniform distribution of proliferating cells. Spheroids were then entrapped in fibrin gels and cultured in serum-free medium and 1 % oxygen. Compared with identical numbers of dissociated MSCs in fibrin gels, spheroids exhibited significantly reduced apoptosis and secreted up to 100-fold more vascular endothelial growth factor. Moreover, fibrin gels containing spheroids and those containing an equivalent number of dissociated cells exhibited similar expression levels of early and late markers of osteogenic differentiation. Thus, MSC spheroids exhibit greater resistance to apoptosis and enhanced proangiogenic potential while maintaining similar osteogenic potential to dissociated MSCs entrapped in a clinically relevant biomaterial, supporting the use of MSC spheroids in cell-based approaches to bone repair.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%