Merck
  • RhoA GTPase-induced ocular hypertension in a rodent model is associated with increased fibrogenic activity in the trabecular meshwork.
RhoA GTPase-induced ocular hypertension in a rodent model is associated with increased fibrogenic activity in the trabecular meshwork.
The American journal of pathology (2014-12-17)
Padmanabhan P Pattabiraman, Tommy Rinkoski, Eric Poeschla, Alan Proia, Pratap Challa, Ponugoti V Rao
ABSTRACT
Ocular hypertension arising from increased resistance to aqueous humor (AH) outflow through the trabecular meshwork is a primary risk factor for open-angle glaucoma, a leading cause of blindness. Ongoing efforts have found little about the molecular and cellular bases of increased resistance to AH outflow through the trabecular meshwork in ocular hypertension patients. To test the hypothesis that dysregulated Rho GTPase signaling and a resulting fibrotic activity within the trabecular meshwork may result in ocular hypertension, we investigated the effects of expressing a constitutively active RhoA GTPase (RhoAV14) in the AH outflow pathway in Sprague-Dawley rats by using lentiviral vector-based gene delivery. Rats expressing RhoAV14 in the iridocorneal angle exhibited a significantly elevated intraocular pressure. Elevated intraocular pressure in the RhoAV14-expressing rats was associated with fibrotic trabecular meshwork and increased levels of F-actin, phosphorylated myosin light chain, α-smooth muscle actin, collagen-1A, and total collagen in the trabecular AH outflow pathway. Most of these changes were ameliorated by topical application of Rho kinase inhibitor. Human autopsy eyes from patients with glaucoma exhibited significant increases in levels of collagen-1A and total collagen in the trabecular AH outflow pathway. Collectively, these observations indicate that increased fibrogenic activity because of dysregulated RhoA GTPase activity in the trabecular AH outflow pathway increases intraocular pressure in a Rho kinase-dependent manner.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O