Merck
  • Determinants of maternal pregnancy one-carbon metabolism and newborn human DNA methylation profiles.
Determinants of maternal pregnancy one-carbon metabolism and newborn human DNA methylation profiles.
Reproduction (Cambridge, England) (2014-11-14)
Nina H van Mil, Marieke I Bouwland-Both, Lisette Stolk, Michael M P J Verbiest, Albert Hofman, Vincent W V Jaddoe, Frank C Verhulst, Paul H C Eilers, Andre G Uitterlinden, Eric A P Steegers, Henning Tiemeier, Régine P M Steegers-Theunissen
ABSTRACT
Maternal one-carbon (1-C) metabolism provides methylgroups for fetal development and programing by DNA methylation as one of the underlying epigenetic mechanisms. We aimed to investigate maternal 1-C biomarkers, folic acid supplement use, and MTHFR C677T genotype as determinants of 1-C metabolism in early pregnancy in association with newborn DNA methylation levels of fetal growth and neurodevelopment candidate genes. The participants were 463 mother-child pairs of Dutch national origin from a large population-based birth cohort in Rotterdam, The Netherlands. In early pregnancy (median 13.0 weeks, 90% range 10.4-17.1), we assessed the maternal folate and homocysteine blood concentrations, folic acid supplement use, and the MTHFR C677T genotype in mothers and newborns. In newborns, DNA methylation was measured in umbilical cord blood white blood cells at 11 regions of the seven genes: NR3C1, DRD4, 5-HTT, IGF2DMR, H19, KCNQ1OT1, and MTHFR. The associations between the 1-C determinants and DNA methylation were examined using linear mixed models. An association was observed between maternal folate deficiency and lower newborn DNA methylation, which attenuated after adjustment for potential confounders. The maternal MTHFR TT genotype was significantly associated with lower DNA methylation. However, maternal homocysteine and folate concentrations, folic acid supplement use, and the MTHFR genotype in the newborn were not associated with newborn DNA methylation. The maternal MTHFR C677T genotype, as a determinant of folate status and 1-C metabolism, is associated with variations in the epigenome of a selection of genes in newborns. Research on the implications of these variations in methylation on gene expression and health is recommended.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%