Merck
  • The indole derivative NecroX-7 improves nonalcoholic steatohepatitis in ob/ob mice through suppression of mitochondrial ROS/RNS and inflammation.
The indole derivative NecroX-7 improves nonalcoholic steatohepatitis in ob/ob mice through suppression of mitochondrial ROS/RNS and inflammation.
Liver international : official journal of the International Association for the Study of the Liver (2014-12-03)
Hyo Kyun Chung, Yong Kyung Kim, Ji-Hoon Park, Min Jeong Ryu, Joon Young Chang, Jung Hwan Hwang, Chul-Ho Lee, Soon-Ha Kim, Hyun Jin Kim, Gi Ryang Kweon, Koon Soon Kim, Minho Shong
ABSTRACT
Nonalcoholic steatohepatitis (NASH) is associated with cirrhosis and hepatocellular carcinoma. Reactive oxygen species (ROS) and reactive nitrogen species (RNS) play key roles in the development of the disease. However, the therapeutic target of NASH has not been fully defined and new treatments are needed. We investigated the protective effects of the antioxidant indole-derived NecroX-7 in a NASH mouse model using leptin-deficient ob/ob and methionine- and choline-deficient (MCD) diet-fed ob/ob mice. Six-week-old male mice were divided into three groups: ob/+ mice, ob/ob mice treated with vehicle and ob/ob mice treated daily with NecroX-7 (20 mg/kg) for 4 weeks. To study the effects of NecroX-7 in a fibrosis model, NASH was induced by feeding ob/ob mice an MCD diet. The effects of NecroX-7 on NASH progression were evaluated using biochemical, histological and molecular markers. NecroX-7-treated ob/ob mice had a marked decrease in serum aspartate aminotransferase and alanine transaminase compared with vehicle-treated controls. Interestingly, hepatic steatosis and lipid peroxidation were significantly improved by NecroX-7 treatment. NecroX-7 inhibited tert-butylhydroperoxide- and H2 O2 -induced mitochondrial ROS/RNS in primary hepatocytes and attenuated mitochondrial dysfunction in vitro and in vivo. Furthermore, NecroX-7-treated mice exhibited fewer infiltrating macrophages and reduced hepatic tumour necrosis factor-alpha expression. Hepatic fibrosis in MCD-fed ob/ob mice was significantly decreased by NecroX-7 treatment. NecroX-7 treatment improved hepatic steatosis and fibrosis in murine NASH models. These effects occurred through the suppression of whole-cell ROS/RNS and inflammatory responses and suggest that NecroX-7 has a potential therapeutic benefit in steatohepatitis.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIS-TRIS
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard