Merck
  • In vitro characterization and in vivo behavior of human nucleus pulposus and annulus fibrosus cells in clinical-grade fibrin and collagen-enriched fibrin gels.
In vitro characterization and in vivo behavior of human nucleus pulposus and annulus fibrosus cells in clinical-grade fibrin and collagen-enriched fibrin gels.
Tissue engineering. Part A (2014-09-23)
Alessandra Colombini, Silvia Lopa, Cristina Ceriani, Arianna B Lovati, Samantha J Croiset, Alessia Di Giancamillo, Giovanni Lombardi, Giuseppe Banfi, Matteo Moretti
ABSTRACT
The intervertebral disc (IVD) presents a limited self-repair ability and cell-based therapies have been suggested to prevent or treat IVD lesions. Fibrin-based scaffolds as cell carriers are promising candidates in IVD tissue engineering, thanks to their ability to be easily delivered into the defect and to adapt to the lesion shape, to support/retain the injected cells into the implantation site and to favor the production of a suitable extracellular matrix (ECM). We evaluated the in vitro and in vivo behavior of human nucleus pulposus (NP) and annulus fibrosus (AF) cells in a clinical-grade collagen-enriched fibrin that has never been tested before for orthopedic applications, comparing it with clinical-grade fibrin. The survival of IVD cells seeded within fibrin or collagen-enriched fibrin and the ECM synthesis were evaluated by biochemical, immunohistochemical, and transcriptional analyses, prior and after subcutaneous implantation of the gels in nude mice. After 28 days of implantation, NP and AF cells were still detectable within explants, produced tissue-specific ECM, and showed a higher content of glycosaminoglycans (GAGs) and type I and II collagen compared to gels before implantation. Both the fibrin gels, enriched or not with collagen, seemed to be suitable for the culture of AF cells, being able to support the homogeneous synthesis of type I collagen, characteristic of the native fibrocartilaginous AF tissue. Differently, fibrin alone was a more suitable matrix for NP culture, supporting the homogeneous deposition of GAGs and type II collagen. In conclusion, our results suggest to combine AF cells with fibrin, enriched or not with collagen, and NP cells with fibrin alone to maintain the typical features of these cell populations, indicating these clinical-grade materials as viable options in cell-based treatments for IVD lesions.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%