Merck
  • A novel folic acid-conjugated TiO₂-SiO₂ photosensitizer for cancer targeting in photodynamic therapy.
A novel folic acid-conjugated TiO₂-SiO₂ photosensitizer for cancer targeting in photodynamic therapy.
Colloids and surfaces. B, Biointerfaces (2014-12-17)
Xiaohui Feng, Shaokun Zhang, Hong Wu, Xia Lou
ABSTRACT
In this paper, a novel folic acid-conjugated silica-coated titanium dioxide (TiO2-SiO2) photosensitizer was synthesized and characterized using various analytical instruments. The photosensitizer was further assessed with regards to its photoreactivity, cellular and hemocompatibility, cell internalization, and phototoxicity. Conjugating folic acid with TiO2-SiO2 has shown a significantly improved compatibility of the nanoparticles with the mouse fibroblast cells (L929) at 24 h. An improved compatibility with the human nasopharyngeal epidermoid cancer (KB) cells was also demonstrated, but to a slightly reduced degree. Enhanced cell internalization was well demonstrated in the TiO2-SiO2 folate nanoparticles. Upon exposure to UV light, TiO2-SiO2 folate nanoparticles maintained a high level photodynamic reactivity and yielded a 38-43% photo-killing of KB cells. The photo-killing effect increased with increasing dosage in the investigated concentration range of 50-100 μg ml(-1).
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Sodium carbonate, powder, 99.999% trace metals basis
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard