Merck
  • P2X1-initiated p38 signalling enhances thromboxane A2-induced platelet secretion and aggregation.
P2X1-initiated p38 signalling enhances thromboxane A2-induced platelet secretion and aggregation.
Thrombosis and haemostasis (2014-03-19)
Z Huang, P Liu, L Zhu, N Li, H Hu
ABSTRACT
ATP released by activated platelets can serve as a positive feedback machinery to amplify platelet responses by activating P2X1 receptors. It has, however, not been defined how P2X1 activities influence thromboxane A2 (TXA2)-stimulated platelet functional responses. Our aim was to elaborate the molecular mechanisms of P2X1 engagements in TXA2-induced platelet secretion and aggregation. P2X1 inhibition by 1 µM NF449 inhibited platelet P-selectin expression induced by a low concentration of the TXA2 analogue U46619 (0.3 µM) (32.0 ± 2.0% vs 43.4 ± 3.0%; n=5; p<0.05). p38 inhibition by SB203580, but not ERK inhibition by U0126, elicited a similar inhibition by NF499. The combination of NF449 and SB203580 provided, however, no additive effects. U46619-induced platelet aggregation was similarly decreased by NF449 and SB203580 alone or in combination, and by P2X1 pre-desensitisation with α,β-Me-ATP. U46619 caused rapid and reversible P2X1-dependent p38 phosphorylation. However, the P2X1-p38 pathway mainly enhanced mild platelet activation by U46619, because α,β-Me-ATP supplementation or p38 blockade had no effect on intense platelet activation induced by a higher concentration of U46619 (3 µM). In conclusion, P2X1 activation, via p38 signalling, potentiates platelet activation initiated by low doses of U46619. Hence, the P2X1-induced p38 signalling promotes more robust platelet activation in response to mild platelet stimuli.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate