Merck
  • Identification of prognosis-related proteins in gingival squamous cell carcinoma by twodimensional gel electrophoresis and mass spectrometry-based proteomics.
Identification of prognosis-related proteins in gingival squamous cell carcinoma by twodimensional gel electrophoresis and mass spectrometry-based proteomics.
Annali italiani di chirurgia (2015-02-26)
Maria Giulia Cristofaro, Domenica Scumaci, Claudia Vincenza Fiumara, Maddalena Di Sanzo, Valeria Zuccalà, Giuseppe Donato, Davide Caruso, Umberto Riccelli, Maria Concetta Faniello, Giovanni Cuda, Francesco Costanzo, Mario Giudice
ABSTRACT
The aim of this study was to identify differentially expressed proteins in oral squamous carcinoma cells that could be potential prognosis-related cancer biomarkers. We compared protein expression patterns from gingival squamous cellc carcinoma (GSCC) tissues and adjacent non-cancerous matched tissues by proteomic analysis using two-dimensional gel electrophoresis coupled to mass spectrometry (2D-PAGE/MS). Seventeen protein spots were found to be over-expressed and eight were under-expressed in cancerous tissue compared to the normal counterpart. Of these, annexin A2 and ezrin were validated by Western blot. We also demonstrated by immunohistochemistry that POSTN is highly expressed in the neoplastic tissues examined. Among the differentially expressed proteins, we focused our attention on Chloride intracellular channel 1 (CLIC1). The 2D-PAGE/MS-based proteomics appears an efficient approach in detecting and identifying differentially expressed proteins that might function as potential biomarkers and/or molecular targets for early cancer diagnosis and prognosis and that might contribute to a innovative therapeutic strategies in GSCC. However, further validation and functional studies are needed to confirm and to support these promising, still preliminary data. Cancer biomarkers, Oral squamous cell carcinoma, Proteomics.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)