Merck
  • Co-delivery of Pirarubicin and Paclitaxel by Human Serum Albumin Nanoparticles to Enhance Antitumor Effect and Reduce Systemic Toxicity in Breast Cancers.
Co-delivery of Pirarubicin and Paclitaxel by Human Serum Albumin Nanoparticles to Enhance Antitumor Effect and Reduce Systemic Toxicity in Breast Cancers.
Molecular pharmaceutics (2015-10-01)
Xiaoli Yi, Xianghong Lian, Jianxia Dong, Zhuoya Wan, Chunyu Xia, Xu Song, Yao Fu, Tao Gong, Zhirong Zhang
ABSTRACT
In our study, we aimed to develop a codelivery nanoparticulate system of pirarubicin (THP) and paclitaxel (PTX) (Co-AN) using human serum albumin to improve the therapeutic effect and reduce systemic toxicities. The prepared Co-AN demonstrated a narrow size distribution around 156.9 ± 3.2 nm (PDI = 0.16 ± 0.02) and high loading efficiency (87.91 ± 2.85% for THP and 80.20 ± 2.21% for PTX) with sustained release profiles. Significantly higher drug accumulation in tumors and decreased distribution in normal tissues were observed for Co-AN in xenograft 4T1 murine breast cancer bearing BALB/c mice. Cytotoxicity test against 4T1 cells in vitro and antitumor assay on 4T1 breast cancer in vivo demonstrated that the antitumor effect of Co-AN was superior to that of the single drug or free combination. Also, Co-AN induced increased apoptosis and G2/M cell cycle arrest against 4T1 cells compared to that of the single drug formulation. Remarkably, Co-AN exhibited significantly lower side effects regarding bone marrow suppression and organ and gastrointestinal toxicities. This human serum albumin-based codelivery system represents a promising platform for combination chemotherapy in breast cancers.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)