Merck
  • Improved DNA microarray detection sensitivity through immobilization of preformed in solution streptavidin/biotinylated oligonucleotide conjugates.
Improved DNA microarray detection sensitivity through immobilization of preformed in solution streptavidin/biotinylated oligonucleotide conjugates.
Colloids and surfaces. B, Biointerfaces (2015-03-26)
E Mavrogiannopoulou, P S Petrou, G Koukouvinos, D Yannoukakos, A Siafaka-Kapadai, K Fornal, K Awsiuk, A Budkowski, S E Kakabakos
ABSTRACT
A novel immobilization approach involving binding of preformed streptavidin/biotinylated oligonucleotide conjugates onto surfaces coated with biotinylated bovine serum albumin is presented. Microarrays prepared according to the proposed method were compared, in terms of detection sensitivity and specificity, with other immobilization schemes employing coupling of biotinylated oligonucleotides onto directly adsorbed surface streptavidin, or sequential coupling of streptavidin and biotinylated oligonucleotides onto a layer of adsorbed biotinylated bovine serum albumin. A comparison was performed employing biotinylated oligonucleotides corresponding to wild- and mutant-type sequences of seven single point mutations of the BRCA1 gene. With respect to the other immobilization protocols, the proposed oligonucleotide immobilization approach offered the highest hybridization signals (at least 5 times higher) and permitted more elaborative washings, thus providing considerably higher discrimination between complimentary and non-complementary DNA sequences for all mutations tested. In addition, the hybridization kinetics were significantly enhanced compared to two other immobilization protocols, permitting PCR sample analysis in less than 40 min. Thus, the proposed oligonucleotide immobilization approach offered improved detection sensitivity and discrimination ability along with considerably reduced analysis time, and it is expected to find wide application in DNA mutation detection.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O