Merck
  • Effects of temperature, pH and NaCl content on in vitro putrescine and cadaverine production through the growth of Serratia marcescens CCM 303.
Effects of temperature, pH and NaCl content on in vitro putrescine and cadaverine production through the growth of Serratia marcescens CCM 303.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2015-09-12)
Zuzana Bubelová, František Buňka, Monika Taťáková, Kateřina Štajnochová, Khatantuul Purevdorj, Leona Buňková
ABSTRACT
The aim of this study was to evaluate the combined effect of temperature (10, 20 and 37°C), pH (4, 5, 6, 7 and 8), and NaCl content (0, 1, 3, 4, 5 and 6% w/v) on the growth and putrescine and cadaverine production of Serratia marcescens CCM 303 under model conditions. The decarboxylase activity of S. marcescens was monitored in broth after cultivation. The cultivation medium was enriched with selected amino acids (ornithine, arginine and lysine; 0.2% w/v each) serving as precursors of biogenic amines. Levels of putrescine and cadaverine in broth were analysed by high-performance liquid chromatography after pre-column derivatisation with o-phthalaldehyde reagent. S. marcescens produced higher amounts of putrescine (up to 2096.8 mg L(-1)) compared to cadaverine content (up to 343.3 mg L(-1)) in all cultivation media. The highest putrescine and cadaverine concentrations were reached during cultivation at 10-20°C, pH 5-7 and NaCl content 1-3% w/v. On the other hand, the highest BAs production of individual cell (recalculated based on a cell; so called "yield factor") was observed at 10°C, pH 4 and salt concentration 3-5% w/v as a response to environmental stress.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
11-(1H-pyrrol-1-yl)undecane-1-thiol, 96%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O