Merck
  • Preferential recognition of isocitrate dehydrogenase by a rabbit monoclonal antibody (ab124797) against the C-terminal peptide of RANKL.
Preferential recognition of isocitrate dehydrogenase by a rabbit monoclonal antibody (ab124797) against the C-terminal peptide of RANKL.
Journal of immunological methods (2015-03-17)
Kazue Terasawa, Anupama R Rajapakshe, Katarzyna A Podyma-Inoue, Chiemi Mishima-Tsumagari, Masaki Yanagishita, Miki Hara-Yokoyama
ABSTRACT
A rabbit monoclonal antibody (Abcam ab124797), with high affinity for a synthetic peptide corresponding to the C-terminal region of the receptor activator of nuclear factor (NF)-κB ligand (RANKL), specifically recognizes a 37 kDa protein by immunoblotting, in good agreement with the molecular mass of RANKL. However, our mass spectroscopy analysis revealed that the protein recognized by the antibody is the α-subunit of NAD(+)-dependent isocitrate dehydrogenase (ICDH), a key Krebs cycle enzyme in mitochondria. Consistently, immunocytochemical staining with the antibody revealed a network organization characteristic of mitochondria, which overlapped with staining by MitoTracker and was lost after the siRNA-mediated downregulation of ICDH. The C-terminal peptide of ICDH contains similar chemical characteristics to that of the RANKL peptide and interacts with the antibody, although the affinity is a hundred times weaker. The present study provides an example of the preferential recognition of a surrogate protein by a rabbit monoclonal antibody.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%