Merck
  • Application of melanin-free ink as a new antioxidative gel enhancer in sardine surimi gel.
Application of melanin-free ink as a new antioxidative gel enhancer in sardine surimi gel.
Journal of the science of food and agriculture (2014-09-27)
Naveen Kumar Vate, Soottawat Benjakul, Tri Winarni Agustini
ABSTRACT
The squid ink that is discarded as waste during processing can be effectively utilised as a gel enhancer in surimi gels, especially those prepared from dark-fleshed fish which have poor gel properties. It also acts as an antioxidant, inhibiting lipid oxidation. This investigation aimed to study the effect of melanin-free ink (MFI) from splendid squid (Loligo formosana) on properties and oxidative stability of surimi gel from sardine (Sardinella albella). MFI (0-0.1 g kg(-1) surimi) increased the breaking force and deformation of sardine surimi gel in a dose-dependent manner (P < 0.05). The addition of MFI had no effect on whiteness of surimi gels (P > 0.05). The expressible moisture content of gels decreased as the levels of MFI increased (P < 0.05). Based on a microstructure study, gel added with MFI at a level of 0.08 g kg(-1) surimi was denser and finer than that of the control (without MFI). Surimi gels with MFI had lower peroxide values, thiobarbituric acid reactive substances, nonanal and 2-decenal. MFI could improve the properties of sardine surimi gel. Additionally, it was able to prevent lipid oxidation in surimi gels during refrigerated storage.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O