Merck
  • Lifelong wheel running exercise and mild caloric restriction attenuate nuclear EndoG in the aging plantaris muscle.
Lifelong wheel running exercise and mild caloric restriction attenuate nuclear EndoG in the aging plantaris muscle.
Experimental gerontology (2015-06-10)
Jong-Hee Kim, Yang Lee, Hyo-Bum Kwak, John M Lawler
ABSTRACT
Apoptosis plays an important role in atrophy and sarcopenia in skeletal muscle. Recent evidence suggests that insufficient heat shock proteins (HSPs) may contribute to apoptosis and muscle wasting. In addition, long-term caloric restriction (CR) and lifelong wheel running exercise (WR) with CR provide significant protection against caspase-dependent apoptosis and sarcopenia. Caspase-independent mediators (endonuclease G: EndoG; apoptosis-inducing factor: AIF) of apoptosis are also linked to muscles wasting with disuse and aging. However, the efficacy of CR and WR with CR to attenuate caspase-independent apoptosis and preserve HSPs in aging skeletal muscle are unknown. Therefore, we tested the hypothesis that CR and WR with CR would ameliorate age-induced elevation of EndoG and AIF while protecting HSP27 and HSP70 levels in the plantaris. Male Fischer-344 rats were divided into 4 groups at 11weeks: ad libitum feeding until 6months (YAL); fed ad libitum until 24months old (OAL); 8%CR to 24months (OCR); WR+8%CR to 24months (OExCR). Nuclear EndoG levels were significantly higher in OAL (+153%) than in YAL, while CR (-38%) and WR with CR (-46%) significantly attenuated age-induced increment in nuclear EndoG. HSP27 (-63%) protein content and phosphorylation at Ser82 (-49%) were significantly lower in OAL than in YAL, while HSP27 protein content was significantly higher in OCR (+136%) and OExCR (+155%) and p-HSP27 (+254%) was significantly higher in OExCR compared with OAL, respectively. In contrast, AIF and HSP70 were unaltered by CR or WR with CR in aging muscle. These data indicate that CR and WR with CR attenuate age-associated upregulation of EndoG translocation in the nucleus, potentially involved with HSP27 signaling.
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture