Merck
  • The impact of intensified training with a high or moderate carbohydrate feeding strategy on resting and exercise-induced oxidative stress.
The impact of intensified training with a high or moderate carbohydrate feeding strategy on resting and exercise-induced oxidative stress.
European journal of applied physiology (2015-03-31)
Alex J Wadley, Sophie C Killer, Ida S Svendsen, Michael Gleeson
ABSTRACT
This study investigated the impact of intensified training (IT) and carbohydrate (CHO) supplementation on resting and exercise-induced oxidative stress. Male cyclists (n = 13, mean ± SD: age 25 ± 6 years; [Formula: see text] 72 ± 5 ml/kg/min) undertook two 9 day periods of endurance-based IT. In a counter-balanced, crossover and double-blinded study design, participants completed IT whilst ingesting high (H-CHO) or moderate (M-CHO) CHO beverages before (H-CHO: 24 g vs. 2 g), during (H-CHO: 60 g/h vs. 20 g/h) and after training sessions (H-CHO: 44 g vs. 10 g). Participants completed fasted performance trials without CHO on days 2, 6 and 10. Blood samples were taken before and immediately after exercise to assess plasma oxidative stress. Resting thiol (-SH) and catalase (CAT) activities decreased following 6 days of IT, independent of CHO condition [-SH (μM oxidised NADPH): H-CHO-14.0 ± 18.8, M-CHO-20.4 ± 20.3 and CAT (nmol/min/ml): H-CHO 12.5 ± 12.5, M-CHO 6.0 ± 4.5; all p < 0.05]. Resting total antioxidant capacity (TAC) was reduced after IT in M-CHO. All exercise bouts elicited significant increases in CAT, TAC, protein carbonylation (PC) and lipid hydroperoxides (LOOH), independent of CHO condition (p < 0.05). The magnitude of increase in PC and LOOH was greater on days 6 and 10 compared to day 2 in both conditions. Short-term IT caused reductions in resting antioxidant capacity in trained cyclists. Exercise-induced increases in PC and LOOH were exaggerated as a result of IT; however, these responses were independent of carbohydrate intake before, during and after the preceding IT sessions.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)