Merck
  • Proteomics for the detection of indirect markers of steroids treatment in bovine muscle.
Proteomics for the detection of indirect markers of steroids treatment in bovine muscle.
Proteomics (2015-03-12)
Roberto Stella, Giancarlo Biancotto, Giorgio Arrigoni, Federica Barrucci, Roberto Angeletti, Peter James
ABSTRACT
Despite the ban by the European Union, anabolic steroids might still be illicitly employed in bovine meat production. The surveillance of misuse of such potentially harmful molecules is necessary to guarantee consumers' health. Analytical methods for drug residue control are based on LC-MS/MS, but their efficacy can be hindered due to undetectable residual concentrations as a result of low-dosage treatments. Screening methods based on the recognition of indirect biological effects of growth promoters' administration, such as the alteration of protein expression, can improve the efficacy of surveillance. The present study was aimed at identifying modifications in the muscle protein expression pattern between bulls treated with an ear implant (Revalor-XS®) containing trenbolone acetate (200 mg) and estradiol (40 mg), and untreated animals. The analysis of skeletal muscle was carried out using a tandem mass tags shotgun proteomics approach. We defined 28 candidate protein markers with a significantly altered expression induced by steroids administration. A subset of 18 candidate markers was validated by SRM and allowed to build a predictive model based on partial least square discriminant analysis. Our findings confirm the effectiveness of the proteomics approach as potential tool to overcome analytical limitations of drug residue monitoring.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerol, FCC, FG