Merck
Synthesis and In Vitro Evaluation of Thiolated Carrageenan.
Journal of pharmaceutical sciences (2015-06-04)
Wongsakorn Suchaoin, Sonja Bonengel, Shah Hussain, Christian W Huck, Benjamin N Ma, Andreas Bernkop-Schnürch
ABSTRACT
The aim of this study was to generate and characterize a thiolated carrageenan. Thiolated carrageenan (carrageenan-SH) was synthesized from kappa (κ)- and iota (ι)-carrageenan by bromine replacement of the hydroxyl moieties followed by substitution to thiol groups using thiourea. Thiolated κ- and ι-carrageenan exhibited 176.57 ± 20.11 and 109.51 ± 18.26 μmol thiol groups per gram polymer, respectively. The resazurin test in Caco-2 cells revealed no toxic effect of both thiolated carrageenans at a concentration below 0.1% (w/v). Regarding efflux pump inhibitory effect, cellular accumulation of multidrug-resistance protein 2 substrate, sulforhodamine 101, was 1.38- and 1.35-fold increased in cells treated with thiolated κ- and ι-carrageenan, respectively. Modification of κ- and ι-carrageenan led to 3.9- and 2.0-fold increase in dynamic viscosity of mucus-thiolated carrageenan mixture within 4 h. Furthermore, residence time of κ- and ι-carrageenan-SH on porcine intestinal mucosa was 6.4- and 1.8-fold prolonged, respectively, as demonstrated by rotating cylinder method, indicating improved mucoadhesive properties. Hence, thiolation of carrageenans led to novel pharmaceutical excipients for various applications.
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Resazurin sodium salt, certified by the BSC
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sulforhodamine 101, Dye content ~95 %
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)