Merck
  • Molecular interactions at the hexadecane/water interface in the presence of surfactants studied with second harmonic generation.
Molecular interactions at the hexadecane/water interface in the presence of surfactants studied with second harmonic generation.
The Journal of chemical physics (2015-06-15)
Yajun Sang, Fangyuan Yang, Shunli Chen, Hongbo Xu, Si Zhang, Qunhui Yuan, Wei Gan
ABSTRACT
It is important to investigate the influence of surfactants on structures and physical/chemical properties of oil/water interfaces. This work reports a second harmonic generation study of the adsorption of malachite green (MG) on the surfaces of oil droplets in a hexadecane/water emulsion in the presence of surfactants including sodium dodecyl sulfate, polyoxyethylene-sorbitan monooleate (Tween80), and cetyltrimethyl ammonium bromide. It is revealed that surfactants with micromolar concentrations notably influence the adsorption of MG at the oil/water interface. Both competition adsorption and charge-charge interactions played very important roles in affecting the adsorption free energy and the surface density of MG at the oil/water interface. The sensitive detection of the changing oil/water interface with the adsorption of surfactants at such low concentrations provides more information for understanding the behavior of these surfactants at the oil/water interface.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O