Merck
  • One-pot liquid-phase exfoliation from graphite to graphene with carbon quantum dots.
One-pot liquid-phase exfoliation from graphite to graphene with carbon quantum dots.
Nanoscale (2015-05-28)
Minghan Xu, Wei Zhang, Zhi Yang, Fan Yu, Yujie Ma, Nantao Hu, Dannong He, Qi Liang, Yanjie Su, Yafei Zhang
ABSTRACT
Carbon quantum dots (CQDs) are novel carbon nanomaterials and are attracting increasing interest due to their good characteristics such as hydrophilicity, chemical stability, quantum yield, small particle sizes, and low cytotoxicity. Herein, we used CQDs as stabilizers and exfoliation agents to exfoliate graphite to graphene in an aqueous medium for the first time. The functions of CQDs are to reduce the surface tension of water to match that of graphite and to make weak interactions (π-π conjugation, hydrophobic force, and the Coulomb attraction) with the graphite surface. Different characterization methods were used to evaluate the presence of layers (<5 layers) of graphene sheets with fewer defects and low oxidation. In the future, CQDs can also be good candidates to exfoliate other two-dimensional materials, such as WS2, BN, MoS2, and g-C3N4, to form two-dimensional heterostructures for a range of possible applications.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Citric acid, 99%