Merck
  • Papillary-cystic pattern is characteristic in mammary analogue secretory carcinomas but is rarely observed in acinic cell carcinomas of the salivary gland.
Papillary-cystic pattern is characteristic in mammary analogue secretory carcinomas but is rarely observed in acinic cell carcinomas of the salivary gland.
Virchows Archiv : an international journal of pathology (2015-05-16)
Min-Shu Hsieh, Yueh-Hung Chou, Shin-Joe Yeh, Yih-Leong Chang
ABSTRACT
Mammary analogue secretory carcinoma (MASC) has a specific ETV6-NTRK3 translocation and morphologically overlaps with acinic cell carcinoma (AciCC). Before the recognition of MASC, in AciCC, four histologic patterns were identified including microcystic, solid, papillary-cystic, and follicular. The aim of this study was to evaluate histologic patterns in these two neoplasms through comprehensive histologic subtyping. Using fluorescence in situ hybridization (FISH), we identified 14 cases of MASC and 21 cases of AciCC. We used comprehensive histologic subtyping to provide a semiquantitive assessment of histologic patterns in each tumor and performed immunohistochemical analyses including S100/vimentin/mammaglobin/DOG1. MASC often presented papillary-cystic patterns without a solid component, previously considered to be one of the four major patterns associated with AciCC. However, in our study, this histologic feature was rarely seen in AciCC and more characteristic of MASC. In aspiration cytology samples, MASC was associated with more cellular atypia. An immunohistochemical panel of S100/mammaglobin/DOG1 was found useful for differential diagnosis. Comprehensive subtyping of histologic patterns is a useful screening method prior to initiation of molecular testing.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)