Merck
  • Quantitative Proteomic Analyses of Molecular Mechanisms Associated with Cytoplasmic Incompatibility in Drosophila melanogaster Induced by Wolbachia.
Quantitative Proteomic Analyses of Molecular Mechanisms Associated with Cytoplasmic Incompatibility in Drosophila melanogaster Induced by Wolbachia.
Journal of proteome research (2015-07-30)
Lin-Ling Yuan, Xiulan Chen, Qiong Zong, Ting Zhao, Jia-Lin Wang, Ya Zheng, Ming Zhang, Zailong Wang, Jeremy C Brownlie, Fuquan Yang, Yu-Feng Wang
ABSTRACT
To investigate the molecular mechanisms of cytoplasmic incompatibility (CI) induced by Wolbachia bacteria in Drosophila melanogaster, we applied an isobaric tags for relative and absolute quantitation (iTRAQ)-based quantitative proteomic assay to identify differentially expressed proteins extracted from spermathecae and seminal receptacles (SSR) of uninfected females mated with either 1-day-old Wolbachia-uninfected (1T) or infected males (1W) or 5-day-old infected males (5W). In total, 1317 proteins were quantified; 83 proteins were identified as having at least a 1.5-fold change in expression when 1W was compared with 1T. Differentially expressed proteins were related to metabolism, immunity, and reproduction. Wolbachia changed the expression of seminal fluid proteins (Sfps). Wolbachia may disrupt the abundance of proteins in SSR by affecting ubiquitin-proteasome-mediated proteolysis. Knocking down two Sfp genes (CG9334 and CG2668) in Wolbachia-free males resulted in significantly lower embryonic hatch rates with a phenotype of chromatin bridges. Wolbachia-infected females may rescue the hatch rates. This suggests that the changed expression of some Sfps may be one of the mechanisms of CI induced by Wolbachia. This study provides a panel of candidate proteins that may be involved in the interaction between Wolbachia and their insect hosts and, through future functional studies, may help to elucidate the underlying mechanisms of Wolbachia-induced CI.
Product Description
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O