Merck
  • Ginsenoside Rh2 inhibits metastasis of glioblastoma multiforme through Akt-regulated MMP13.
Ginsenoside Rh2 inhibits metastasis of glioblastoma multiforme through Akt-regulated MMP13.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-04-04)
Ning Guan, Xiaochuan Huo, Zhenxing Zhang, Shoudan Zhang, Junsheng Luo, Wenshi Guo
ABSTRACT
Glioblastoma multiforme (GBM) is the most malignant type of primary brain tumor. Although the growth of the tumor cells in a relatively closed space may partially account for its malignancy, highly invasive nature of glioblastoma cells has been suggested to be the main reason for the failure of current therapeutic approaches. Ginsenoside Rh2 (GRh2) has recently been shown to significantly suppress the growth and survival of GBM through inhibiting epidermal growth factor receptor signaling, whereas its effects on the invasion and metastasis have not been examined. Here, we showed that GRh2 dose-dependently decreased GBM cell invasiveness in both scratch wound healing assay and Transwell cell migration assay. Moreover, the inhibitory effects of GRh2 on cell migration seemed to be conducted through decreased expression of matrix metalloproteinase (MMP)-13. Furthermore, using specific inhibitors, we found that GRh2 inhibited MMP13 through PI3k/Akt signaling pathway. Finally, high MMP13 levels were detected in GBM specimen from the patients. Together, these data suggest that GRh2 may suppress GBM migration through inhibiting Akt-mediated MMP13 activation. Thus, our data highlight a previous unappreciated role for GRh2 in suppressing GBM cell metastasis.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Glycerin, meets USP testing specifications