Merck
  • Ameliorating Effect of Akebia quinata Fruit Extracts on Skin Aging Induced by Advanced Glycation End Products.
Ameliorating Effect of Akebia quinata Fruit Extracts on Skin Aging Induced by Advanced Glycation End Products.
Nutrients (2015-11-17)
Seoungwoo Shin, Dahee Son, Minkyung Kim, Seungjun Lee, Kyung-Baeg Roh, Dehun Ryu, Jongsung Lee, Eunsun Jung, Deokhoon Park
ABSTRACT
The accumulation of free radicals and advanced glycation end products (AGEs) in the skin plays a very important role in skin aging. Both are known to interact with each other. Therefore, natural compounds or extracts that possess both antioxidant and antiglycation activities might have great antiageing potential. Akebia quinata fruit extract (AQFE) has been used to treat urinary tract inflammatory disease in traditional Korean and Chinese medicines. In the present study, AQFE was demonstrated to possess antioxidant and antiglycation activity. AQFE protects human dermal fibroblasts (HDFs) from oxidative stress and inhibits cellular senescence induced by oxidative stress. We also found that AQFE inhibits glycation reaction between BSA and glucose. The antiglycation activity of AQFE was dose-dependent. In addition, the antiglycation activity of AQFE was confirmed in a human skin explant model. AQFE reduced CML expression and stimulated fibrillin-1 expression in comparison to the methyglyoxal treatment. In addition, the possibility of the extract as an anti-skin aging agent has also been clinically validated. Our analysis of the crow's feet wrinkle showed that there was a decrease in the depth of deep furrows in RI treated with AQFE cream over an eight-week period. The overall results suggest that AQFE may work as an anti-skin aging agent by preventing oxidative stress and other complications associated with AGEs formation.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%