Merck
  • Antibacterial activity of fluoride compounds and herbal toothpastes on Streptococcus mutans: an in vitro study.
Antibacterial activity of fluoride compounds and herbal toothpastes on Streptococcus mutans: an in vitro study.
Australian dental journal (2014-10-29)
J P Randall, W K Seow, L J Walsh
ABSTRACT
Streptococcus mutans is an important bacterial species implicated in dental caries. This laboratory study compared the antimicrobial activity of a number of fluoride containing and herbal dentifrices and their components against S. mutans. An agar diffusion method was used with Mueller-Hinton agar. Wells were filled with either 10 commercial fluoride or 6 herbal dentifrices, or with solutions of various fluoride compounds, sodium lauryl sulphate, sodium benzoate, chlorhexidine digluconate or triclosan. Diameters of zones of bacterial growth inhibition surrounding the wells were measured using a micrometer. Significant differences were found for growth inhibition between the 10 fluoridated dentifrices (p < 0.0001), with Colgate Total having the greatest effect. There was not a direct correlation with fluoride type or fluoride concentration. The antibacterial activities of the 6 herbal toothpastes varied, with Herbal Fresh being the strongest. Sodium lauryl sulphate showed strong antimicrobial activity against S. mutans at the levels used in dentifrices. Antimicrobial activity of commercial dentifrices against S. mutans may be exerted by components other than fluoride. Ingredients such as triclosan and sodium lauryl sulphate have larger antimicrobial effects than fluorides in this model.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Chlorhexidine diacetate salt hydrate, bis(biguanide) antimicrobial
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Tin(II) fluoride, 99%
Sigma-Aldrich
Sodium fluorophosphate, 95%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Sodium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O