Merck
  • Guggulsterone attenuates cerulein-induced acute pancreatitis via inhibition of ERK and JNK activation.
Guggulsterone attenuates cerulein-induced acute pancreatitis via inhibition of ERK and JNK activation.
International immunopharmacology (2015-04-07)
Dong-Goo Kim, Gi-Sang Bae, Sun-Bok Choi, Il-Joo Jo, Joon-Yeon Shin, Sung-Kon Lee, Myoung-Jin Kim, Min-Jun Kim, Hyun-Woo Jeong, Chang-Min Choi, Seung-Hee Seo, Gab-Chul Choo, Sang-Wan Seo, Ho-Joon Song, Sung-Joo Park
ABSTRACT
Guggulsterone (GS), a plant steroid and a compound found at high levels in Commiphora myrrha, exhibits anti-inflammatory, anti-cancer, and cholesterol-lowering effects. However, the potential of GS to ameliorate acute pancreatitis (AP) is unknown. The aim of this study was to evaluate the effects of GS on cerulein-induced AP. AP was induced by intraperitoneally injecting supramaximal concentrations of the stable cholecystokinin analog cerulein (50 μg/kg) hourly for 6 h. In the GS-treated group, GS was administered intraperitoneally (10, 25, or 50mg/kg) 1 h before the first cerulein injection. Mice were sacrificed 6 h after the final cerulein injection. Blood samples were collected to measure serum lipase levels and evaluate cytokine production. The pancreas and lung were rapidly removed for morphologic and histological examinations, flow cytometry analysis, myeloperoxidase (MPO) assay, and real-time reverse transcription-polymerase chain reaction analysis. Pre-treatment with GS attenuated cerulein-induced histological damage, reduced pancreas weight/body weight ratio, decreased serum lipase levels, inhibited infiltrations of macrophages and neutrophils, and suppressed cytokine production. Additionally, GS treatment suppressed the activation of extracellular signal-regulated protein kinase (ERK) and c-Jun N-terminal kinase (JNK) in the pancreas in cerulein-induced pancreatitis. In conclusion, our results suggest that GS attenuates AP via deactivation of ERK and JNK.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
(E)-Guggulsterone, ≥95% (HPLC), powder
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O